Balloon Helicopter Assembly | Avantor
Balloon Helicopter Assembly
Supplier:   American Scientific
Balloon Helicopter Assembly
Certificates
Balloon Helicopter Assembly
Balloon Helicopter Assembly
2 Options Available Below
Product Details & Documents
Effectively illustrate Newton’s Third Law with this exciting and simple discrepant event.
  • Uses 9" round balloons
  • Class pack includes 25 kits and air pump
  • Assorted colors
The product allows you to conduct a fun demonstration that is great for younger kids learning about science. It makes a great addition to science classrooms!
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Balloon helicopter assembly, single
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3.08
    $3.08
    Each
       $3.08
    Supplier Number: 7030-00
  • Balloon helicopter assembly, class pack of 25
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 12
    Kit of 12 - $32.39
    $32.39
    Kit of 12
       $32.39
    Supplier Number: 7030-03