Magleads | Avantor
Magleads
Supplier:   American Scientific
Magleads
Certificates
Magleads
3 Options Available Below
Product Details & Documents
Simply the easiest and best jumper wires you can have in your classroom.
  • The 4 mm diameter magnets work in all the places that standard clips work
  • No need for an adapter to connect wires to your battery
  • No more fighting with Fahnestock or alligator clips - just click them together!
Instantly make working electrical circuits with these ingenious leads, each fitted with a tiny powerful magnet at either end. Place a couple near to a battery case and they connect by themselves. Then, place them next to a bulb holder connector and you have your first working circuit!
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Longueur
    Availability
    Price
  • Magleads, pack of 10
    Clear
    12"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $38.39
    $38.39
    Pack of 10
       $38.39
    Supplier Number: ML300C
  • Magleads, pack of 10
    Black
    12"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $39.82
    $39.82
    Pack of 10
       $39.82
    Supplier Number: ML300B
  • Magleads, pack of 10
    Red
    12"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $39.82
    $39.82
    Pack of 10
       $39.82
    Supplier Number: ML300R