Confining Clay Layer Mix
Confining clay layer mix, 1.5 lbs.
Catalog # 470038-606
Supplier:   Avantor
Confining Clay Layer Mix
Catalog # 470038-606
Supplier:  Avantor
Confining Clay Layer Mix
Confining Clay Layer Mix
Confining clay layer mix, 1.5 lbs.
Price
$26.05
Log in pour la disponibilité
Pack of 1
Product Details & Documents
Mix designed to experiment with confined clay.
  • Mix of fine black sand and pure monmorillonite powder
  • Housed in a 16 oz. jar with screw cap
Spécifications
  • Désignation:
    Confining clay layer mix, 1.5 lbs.