Toothed Forceps | Avantor
Toothed Forceps
Supplier:   AFTAB AHMED SE
Toothed Forceps
Supplier:  AFTAB AHMED SE
Certificates
Toothed Forceps
3 Options Available Below
Product Details & Documents
Toothed grip for a secure hold.
  • Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all have a toothed jaw to provide a very secure grip on tough or slippery tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Adison brown
    Stainless steel
    4.5"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.10
    $11.10
    Log in pour la disponibilité
    Box of 12
    Box of 12 - $126.35
    $126.35
    Each
       $11.10
    Box of 12
       $126.35
    Supplier Number: 15-103
  • Tissue forceps
    Stainless steel
    4.5"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1.40
    $1.40
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $69.40
    $69.40
    Each
       $1.40
    Box of 50
       $69.40
    Supplier Number: 14-1015
  • Gilles forceps
    Stainless steel
    4.5"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9.75
    $9.75
    Log in pour la disponibilité
    Box of 12
    Box of 12 - $112.10
    $112.10
    Each
       $9.75
    Box of 12
       $112.10
    Supplier Number: 14-1017