Serrated Forceps
À propos de cet article
Serrated grips for less tissue damage.
- Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all feature a serrated jaw to provide a secure grip without causing significant tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
13 Options Available Below
Product Details & Documents
Serrated grips for less tissue damage.
- Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all feature a serrated jaw to provide a secure grip without causing significant tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Fine tip
Informations sur le produit
- MatièreLongueurTip StyleAvailabilityPrice
- Stainless steel4.5"BentEach$2.30Box of 50$107.60Supplier Number: 14-1009
- Stainless steel4.5"StraightEach$1.45Box of 50$70.50Supplier Number: 14-1004
- Stainless steel4.5"CurvedEach$4.06Box of 50$203.00Supplier Number: 14-0531
- Stainless steel4.5"StraightEach$1.45Box of 50$70.50Supplier Number: 14-0541
- Stainless steel5"CurvedEach$1.40Box of 50$69.40Supplier Number: 14-0532
Medium tip
Informations sur le produit
- MatièreLongueurTip StyleAvailabilityPrice
- Stainless steel4.5"StraightEach$1.45Box of 50$70.50Supplier Number: 14-1001
- Stainless steel4.5"CurvedEach$6.10Box of 50$294.35Supplier Number: 14-1003
- Stainless steel4.5"StraightEach$6.30Box of 12$71.95Supplier Number: 14-1000
- Stainless steel4.5"BentEach$4.13Box of 50$206.50Supplier Number: 15-109
- Stainless steel8"StraightEach$9.25Box of 12$104.90Supplier Number: 14-1005
Broad tip
Informations sur le produit
- MatièreLongueurTip StyleAvailabilityPrice
- Stainless steel4.5"StraightEach$1.40Box of 50$69.75Supplier Number: 14-0999
- Stainless steel6"StraightEach$7.55Box of 12$86.00Supplier Number: 15-111
- Stainless steel8"StraightEach$14.70Box of 12$167.30Supplier Number: 14-1007