Hemostatic Forceps
Hemostatic Forceps
Supplier:   AFTAB AHMED SE
Hemostatic Forceps
Supplier:  AFTAB AHMED SE
Certificates
Hemostatic Forceps
Hemostatic Forceps
1 Options Available Below
Product Details & Documents
Lock for a secure hold.
  • Stainless steel
  • Serrated jaws
Hemostatic forceps are a locking forceps, with thumb loops that operate much like scissors. Often referred to as clamps, these have a serrated jaw to provide a secure hold while minimizing tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Product Information
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Halstead mosquito clamp
    Stainless steel
    5"
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $8.25
    $8.25
    Log in for availability
    Box of 12
    Box of 12 - $93.70
    $93.70
    Each
       $8.25
    Box of 12
       $93.70
    Supplier Number: 14-0551
  • Hemostatic forceps
    Stainless steel
    5.5"
    Curved
    Discontinued
    Supplier Number: 20-105