Special Use Forceps
Special Use Forceps
Special Use Forceps
Special Use Forceps
Specialty forceps for student use.
  • Stainless steel
  • A variety of options available
One set of forceps isn't like every other. This selection provides options for many users. Tweezers are a basic sharp point forceps with a very short blade length that makes the ends very easy to control and provides a stronger grip.
Cover slipping forceps are designed with a broader smooth tip that is bent at an angle to improve applying a coverslip to a microscope slide.
Microdissection forceps are similar to a watchmakers forceps with a lighter weight blade, requiring less force to close. Easy grip forceps have an enlarged grip loop part-way down each blade to provide additional stability and ease of closure for a narrow blade.
Finally the self closing forceps act in a reverse motion to most. Squeezing opens the jaws, and they close when released. These are easier to use if long-term holding is required.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Désignation
    Matière
    Longueur
    Tip Style
  • Tweezers
    Stainless steel
    4.5"
    Straight
    Each
    Each - $6.15
    $6.15
    Box of 12
    Box of 12 - $69.75
    $69.75
    Each
       $6.15
    Box of 12
       $69.75
     : 14-0512
  • Self closing
    Stainless steel
    4.5"
    Straight
    Each
    Each - $6.90
    $6.90
    Box of 12
    Box of 12 - $78.15
    $78.15
    Each
       $6.90
    Box of 12
       $78.15
     : 15-122
  • Microdissection X-fine
    Stainless steel
    Curved
    Each
    Each - $2.65
    $2.65
    Box of 50
    Box of 50 - $125.70
    $125.70
    Each
       $2.65
    Box of 50
       $125.70
     : 14-1010
  • Ez-grip fine point
    Stainless steel
    Straight
     : 14-0590
  • Cover glass forceps
    Stainless steel
    Bent
    Each
    Each - $7.05
    $7.05
    Box of 12
    Box of 12 - $79.15
    $79.15
    Each
       $7.05
    Box of 12
       $79.15
     : 15-TW088
  • Featherweight fine tip
    Stainless steel
    Each
    Each - $13.75
    $13.75
    Box of 12
    Box of 12 - $156.90
    $156.90
    Each
       $13.75
    Box of 12
       $156.90
     : 14-0520