Watchmaker Forceps
Supplier:   AFTAB AHMED SE
Watchmaker Forceps
Supplier:  AFTAB AHMED SE
Certificates
Watchmaker Forceps
Watchmaker Forceps
2 Options Available Below
Product Details & Documents
Ultra-fine point forceps.
  • Stainless steel
Extremely fine tips and heavier handles allow for the manipulation of very fine tissue samples with a secure hold.
These forceps have a smooth finish inside the jaws, to ensure a minimum of tissue damage.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Matière
    Longueur
    Tip Style
    Availability
    Price
  • Stainless steel
    4"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 9,65 $
    9,65 $
    Log in pour la disponibilité
    Box of 12
    Box of 12 - 110,35 $
    110,35 $
    Each
       9,65 $
    Box of 12
       110,35 $
    Supplier Number: 14-1400
  • Stainless steel
    4.25"
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 9,00 $
    9,00 $
    Log in pour la disponibilité
    Box of 12
    Box of 12 - 103,20 $
    103,20 $
    Each
       9,00 $
    Box of 12
       103,20 $
    Supplier Number: 14-0505