Dissection Scissors
Dissection Scissors
Dissection Scissors
Dissection Scissors
Basic dissection for students.
  • Stainless steel
Basic dissection scissors are strong, durable, and have shorter handles for good control. A good option for inexperienced students, these have a slightly blunted pointed blade set to also minimize the chance for accidental pokes.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Désignation
    Longueur
    Matière
    Blade Type
  • Economy student scissors
    4.5"
    Stainless steel
    Sharp/sharp
    Each
    Each - $7.15
    $7.15
    Box of 12
    Box of 12 - $79.95
    $79.95
    Each
       $7.15
    Box of 12
       $79.95
     : 14-0525
  • Student scissors
    4.5"
    Stainless steel
    Sharp/sharp
    Each
    Each - $8.35
    $8.35
    Box of 12
    Box of 12 - $94.15
    $94.15
    Each
       $8.35
    Box of 12
       $94.15
     : 14-0600
  • Fine tip
    4.5"
    Stainless steel
    Sharp/sharp
    Each
    Each - $6.35
    $6.35
    Box of 12
    Box of 12 - $71.40
    $71.40
    Each
       $6.35
    Box of 12
       $71.40
     : 14-0980
  • Scissors straight blade
    5.5"
    Stainless steel
    Sharp/sharp
    Each
    Each - $12.55
    $12.55
    Box of 12
    Box of 12 - $143.05
    $143.05
    Each
       $12.55
    Box of 12
       $143.05
     : 10-917
  • Mayo scissors
    5.5"
    Stainless steel
    Sharp/sharp
    Each
    Each - $8.35
    $8.35
    Box of 12
    Box of 12 - $94.15
    $94.15
    Each
       $8.35
    Box of 12
       $94.15
     : 10-916