Iris Scissors
Iris Scissors
Iris Scissors
Iris Scissors
Extremely fine scissors.
  • Stainless steel
Double sharp points, a very fine blade thickness, and shorter handles make Iris scissors the easiest to control in very tight locations. The blades will very cleanly cut even very fine tissue.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Désignation
    Longueur
    Matière
    Blade Type
  • Iris scissors
    4.5"
    Tungsten carbide
    Sharp/sharp
    Each
    Each - $38.35
    $38.35
    Box of 12
    Box of 12 - $439.10
    $439.10
    Each
       $38.35
    Box of 12
       $439.10
     : 10-901
  • Closed shank
    4.5"
    Stainless steel
    Sharp/sharp
    Each
    Each - $8.10
    $8.10
    Box of 12
    Box of 12 - $93.00
    $93.00
    Each
       $8.10
    Box of 12
       $93.00
     : 14-0601
  • Micro iris
    4.75"
    Stainless steel
    Sharp/sharp
    Each
    Each - $24.80
    $24.80
    Box of 12
    Box of 12 - $284.15
    $284.15
    Each
       $24.80
    Box of 12
       $284.15
     : 14-0976