Dissection Needles | Avantor
Dissection Needles
Supplier:   AFTAB AHMED SE
Dissection Needles
Supplier:  AFTAB AHMED SE
Certificates
Dissection Needles
Dissection Needles
2 Options Available Below
Product Details & Documents
Sharp point needles.
  • Fine point teasing needles
  • Straight or angled tips
Very sharp points on these needles make tissue separation fast and easy. Used mostly to break apart muscle tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Straight teasing needle
    Stainless steel
    5.5"
    Sharp
    Discontinued
    Supplier Number: 14-1033
  • Webster needle holder
    Stainless steel
    6.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 8,25 $
    8,25 $
    Log in pour la disponibilité
    Box of 12
    Box of 12 - 93,70 $
    93,70 $
    Each
       8,25 $
    Box of 12
       93,70 $
    Supplier Number: 20-103
  • Needle for needle holder
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 5,05 $
    5,05 $
    Log in pour la disponibilité
    Box of 100
    Box of 100 - 469,75 $
    469,75 $
    Each
       5,05 $
    Box of 100
       469,75 $
    Supplier Number: 14-0633