Dissection Probes
Dissection Probes
Supplier:   AFTAB AHMED SE
Dissection Probes
Supplier:  AFTAB AHMED SE
Certificates
Dissection Probes
Dissection Probes
3 Options Available Below
Product Details & Documents
Easily tease up fine tissues.
  • Blunt tips prevent accidental tissue damage
  • Stainless steel
Probes provide an easy way to lift embedded tissue, or track the path of nerves and vessels.
The spear point is sharp to allow controlled removal of connective tissue, while the blunt probes allow for easily lifting tissue and elevating.
Blunt probes are also very good for breaking through loose connective tissue without damaging other tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Angular probe and seeker
    Stainless steel
    6"
    Blunt/blade
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5.40
    $5.40
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $129.05
    $129.05
    Each
       $5.40
    Box of 25
       $129.05
    Supplier Number: 14-0958
  • Flexible probe
    Stainless steel
    6"
    Blunt/notched
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4.10
    $4.10
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $392.55
    $392.55
    Each
       $4.10
    Box of 100
       $392.55
    Supplier Number: 14-0959
  • Angled probe and seeker
    Stainless steel
    6"
    Blunt
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5.45
    $5.45
    Log in pour la disponibilité
    Box of 25
    Box of 25 - $130.05
    $130.05
    Each
       $5.45
    Box of 25
       $130.05
    Supplier Number: 14-0950