Dissection Knives
Supplier:   AFTAB AHMED SE
Dissection Knives
Supplier:  AFTAB AHMED SE
Certificates
Dissection Knives
Dissection Knives
3 Options Available Below
Product Details & Documents
Great for rough dissection.
  • Stainless steel construction
  • Fixed blades for safety
More durable than scalpels, dissection knives are great for rough dissection, rapid fleshing, or dealing with tough tissues. The basic fleshing knife has a shorter blade and is easily controlled.
The Amputation knife has an extended blade for long smooth cuts through soft tissues. Use the cartilage knife, which has a scalpel form, to easily reach into tight areas to cut tough tissue.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Availability
    Price
  • Cartilage knife
    Stainless steel
    6.5"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10.50
    $10.50
    Log in pour la disponibilité
    Box of 12
    Box of 12 - $118.05
    $118.05
    Each
       $10.50
    Box of 12
       $118.05
    Supplier Number: 14-0930
  • Fleshing knife
    Stainless steel
    7"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $19.20
    $19.20
    Log in pour la disponibilité
    Box of 12
    Box of 12 - $220.00
    $220.00
    Each
       $19.20
    Box of 12
       $220.00
    Supplier Number: 14-1190
  • Liston amputation knife
    Stainless steel
    8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $38.90
    $38.90
    Log in pour la disponibilité
    Box of 6
    Box of 6 - $221.70
    $221.70
    Each
       $38.90
    Box of 6
       $221.70
    Supplier Number: 14-0969