Magnifying Forceps
Forceps magnifying glass, Stainless steel, 4.5"
Catalog # 470334-798
Supplier: AFTAB AHMED SE
Fournisseur #: 10-PM1482
Jump to:
Product Details & Documents
Great for insect pinning.
- Easily grasp small items
- Stainless steel
A simple set of fine point forceps with an attached 4× magnifier. Ideal for the manipulation of small items, or convenient for use with hard to identify structures.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Forceps magnifying glass
- Longueur:4.5"
- Matière:Stainless steel