Bone Shears | Avantor
Bone Shears
Supplier:   AFTAB AHMED SE
Bone Shears
Supplier:  AFTAB AHMED SE
Certificates
Bone Shears
3 Options Available Below
Product Details & Documents
Easily snip through bones.
  • Stainless steel
  • Flush cutting face
  • Short jaws for additional force
Used to easily cut through tough tissue or bone during dissection. These bone cutting shears combine longer grips and shorter heads to make cuts easier. The cutter has a beveled edge on one side for easy flush cutting.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Matière
    Availability
    Price
  • Small bone cutting shears
    5"
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $76.75
    $76.75
    Log in pour la disponibilité
    Box of 6
    Box of 6 - $436.30
    $436.30
    Each
       $76.75
    Box of 6
       $436.30
    Supplier Number: 14-1110
  • Bone cutter, fine point
    5.5"
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $51.75
    $51.75
    Log in pour la disponibilité
    Box of 6
    Box of 6 - $294.85
    $294.85
    Each
       $51.75
    Box of 6
       $294.85
    Supplier Number: 14-0995
  • Large bone cutting shears
    6.5"
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $79.30
    $79.30
    Log in pour la disponibilité
    Box of 6
    Box of 6 - $451.80
    $451.80
    Each
       $79.30
    Box of 6
       $451.80
    Supplier Number: 14-0997