À propos de cet article
Codeable, 3D-printed robotic hand for interactive STEM learning.
- Educational focus on medical innovation
- Easy to use and assemble: features a solderless breadboard and simple components
- Powered by Arduino Uno
- Optional online courses foster creativity and problem-solving in a tech-focused educational environment
The robotics hand standard kit is perfect for learning about medical innovation and bionics. It includes everything needed to build a robotic hand: a 3D-printed hand model, five micro servo motors to move the hand, servo caps for connecting the motors, hookup wires for connections, an Arduino Uno for control, a USB cable for connecting to a computer, an extra USB wire, two types of capacitors for circuit stability, a voltage regulator, and a solderless breadboard for easy circuit building. This kit is great for students and hobbyists interested in robotics and medical technology.
Upgrade to the special Design Entrepreneurship and STEM Skills (DESS) courses, with extra support. These courses combine design, entrepreneurship, and STEM education to boost creativity and innovation. It's designed to develop top-notch problem-solving skills, preparing students for a tech-focused future and enabling them to tackle complex challenges effectively.