PicoSolutions 4-Axis Robotic Arm
À propos de cet article
4-axis educational robotic arm: precision learning tool.
- 4 precision axes: offers a broad range of motion for versatile tasks from simple placements to complex maneuvers
- User-friendly coding: works seamlessly with major block-based coding platforms like Scratch and Blockly
- Compact design is ideal for desktop use in classrooms, homes, or labs; maximize space without sacrificing functionality
- Easily customizable with add-ons like sensors and attachments to grow with user skills and demands
- Supplied with a wealth of tutorials and learning materials to support users of all experience levels
Introducing the mini educational 4-axis robotic arm – a gateway to the world of robotics and coding. This compact yet powerful robotic arm is designed to teach users of all ages about the fundamentals of robotics, programming, and automation. Standing as a marvel of modern engineering, the Mini 4-Axis Robotic Arm boasts four precision-controlled axes, allowing for a wide range of movement and functionality. From simple pick-and-place tasks to more complex manipulations, this versatile arm can handle it all with ease.
What truly sets this robotic arm apart is its intuitive block coding interface. No prior coding experience is required – simply drag and drop blocks of code to command the arm's movements and actions. With a user-friendly interface, beginners can quickly grasp the basics of programming while seasoned enthusiasts can delve into more advanced concepts. Whether you're a student exploring STEM subjects, an educator looking to enhance classroom learning, or a hobbyist eager to dive into robotics, the Mini Educational 4-axis robotic arm offers endless possibilities for experimentation and discovery.
Upgrade to the special Design Entrepreneurship and STEM Skills (DESS) courses, with extra support. These courses combines design, entrepreneurship, and STEM education to boost creativity and innovation. It's designed to develop top-notch problem-solving skills, preparing students for a tech-focused future and enabling them to tackle complex challenges effectively.
5 Options Available Below
Product Details & Documents
4-axis educational robotic arm: precision learning tool.
- 4 precision axes: offers a broad range of motion for versatile tasks from simple placements to complex maneuvers
- User-friendly coding: works seamlessly with major block-based coding platforms like Scratch and Blockly
- Compact design is ideal for desktop use in classrooms, homes, or labs; maximize space without sacrificing functionality
- Easily customizable with add-ons like sensors and attachments to grow with user skills and demands
- Supplied with a wealth of tutorials and learning materials to support users of all experience levels
Introducing the mini educational 4-axis robotic arm – a gateway to the world of robotics and coding. This compact yet powerful robotic arm is designed to teach users of all ages about the fundamentals of robotics, programming, and automation. Standing as a marvel of modern engineering, the Mini 4-Axis Robotic Arm boasts four precision-controlled axes, allowing for a wide range of movement and functionality. From simple pick-and-place tasks to more complex manipulations, this versatile arm can handle it all with ease.
What truly sets this robotic arm apart is its intuitive block coding interface. No prior coding experience is required – simply drag and drop blocks of code to command the arm's movements and actions. With a user-friendly interface, beginners can quickly grasp the basics of programming while seasoned enthusiasts can delve into more advanced concepts. Whether you're a student exploring STEM subjects, an educator looking to enhance classroom learning, or a hobbyist eager to dive into robotics, the Mini Educational 4-axis robotic arm offers endless possibilities for experimentation and discovery.
Upgrade to the special Design Entrepreneurship and STEM Skills (DESS) courses, with extra support. These courses combines design, entrepreneurship, and STEM education to boost creativity and innovation. It's designed to develop top-notch problem-solving skills, preparing students for a tech-focused future and enabling them to tackle complex challenges effectively.
Product Family Options
Informations sur le produit
- DésignationTailleAvailabilityPrice
- Supplier Number: PS-4000-01
- Supplier Number: PS-4000-01-PK
- 4-axis robotic arm + D.E.S.S. course 101 + 1.5 hours support12H×11 W×11"DLog in pour la disponibilitéEach$2,607.00Supplier Number: PS-4000-01-D1
- 4-axis robotic arm + D.E.S.S. courses 101 and 102 + 1.5 hours support12H×11 W×11"DLog in pour la disponibilitéEach$2,765.00Supplier Number: PS-4000-01-D2
- 4-axis robotic arm + D.E.S.S. courses complete + 1.5 hours support12H×11 W×11"DLog in pour la disponibilitéEach$2,844.00Supplier Number: PS-4000-01-D3