Combitips® advanced Positive Displacement Pipette Tips, PCR Clean
   Sustainable Options Available
Supplier:   Eppendorf
Combitips® advanced Positive Displacement Pipette Tips, PCR Clean
Supplier:  Eppendorf
   Sustainable Options Available
Certificates
Combitips® advanced Positive Displacement Pipette Tips, PCR Clean
Combitips® advanced Positive Displacement Pipette Tips, PCR Clean
9 Options Available Below
Product Details & Documents
Eppendorf Combitips® advanced in 'PCR clean' grade offer superior positive displacement pipetting, ensuring precise handling of sensitive liquids in PCR and molecular biology, with contamination-free, ergonomic design for reliable lab results.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0.1 to 50 ml) offer a maximum range of dispensing volumes with increments as low as 100 nl
  • Certified to be free from human DNA, DNase, RNase, and PCR inhibitors that could adversely affect polymerase chain reactions
  • Lot-specific certificates for Eppendorf consumables in 'PCR clean' quality are available
  • Elongated Combitips® advanced in volumes 2.5, 5.0, and 10 ml will reach the bottom of most common laboratory vessels
  • Combitips® advanced are offered in five different purity grades to support a range of applications
  • Appropriate purity grade for each application: Eppendorf Quality, PCR clean, Forensic DNA Grade, sterile, or Biopur®
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability
The tip of choice when working with Repeater®/Multipette® multi-dispenser pipettes from Eppendorf:
Thanks to their revolutionary design, Combitips® advanced dispenser tips provide many benefits for work in a modern laboratory. With an integrated piston, they function according to the positive displacement principle and ensure that the correct volume of liquid is dispensed regardless of the physical properties of the liquid (e.g., viscosity, volatility, density, temperature). Achieve high-precision dispensing, independent of the flow characteristics of the liquid, and safely work with radioactive or toxic materials due to the hermetically sealed piston that prevents aerosol contamination.
Eppendorf pioneered the Repeater®/Combitips® multi-dispenser system in 1978 and improved it several times since. It was the very first pipetting system allowing users to fill the tip once and dispense multiple times. The concept remains a driver for efficient liquid handling and a staple in many labs working with plates or series of tubes. In 2023, the Combitips® advanced pipette tip line-up has been certified with ACT® Environmental Impact Factor Labels by the My Green Lab® non-profit organization, making it the ideal choice for labs seeking more sustainable products.
 : ACT® label and lot-specific certficates available for the purity grade PCR clean can be downloaded from the Eppendorf website.
 : Combitips® advanced are designed for use with Eppendorf Repeater®/Multipette® multi-dispensers only.
Specifications for Product Family
Specifications
  • Catalog No:
  • 89510-208
  • CA11033-306
  • 89510-210
  • CA11033-308
  • 89510-212
  • CA11033-310
  • 89510-214
  • CA11033-312
  • 89510-216
  • CA11033-314
  • 89510-218
  • CA11033-316
  • 89510-220
  • CA11033-318
  • 89510-222
  • CA11033-320
  • 89511-338
  • CA11033-322
  • Environmentally Preferable:
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
  • Renewable Energy Certificate, ISO 14001, ACT Label
Specifications
Product Family Options
Product Information
  • Volume
    Color Code
    Packed
    Availability
    Price
  • 0.1 ml
    White
    100 pieces (4 reclosable bags×25 pieces)
    Discontinued
    Supplier Number: 0030089766
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89510-208
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 0.1 ml
    White
    100 pieces (4 reclosable bags×25 pieces)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $343.00
    $343.00
    Pack of 100
       $343.00
    Supplier Number: 0030089766
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-306
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 0.2 ml
    Light blue
    100 pieces (4 reclosable bags×25 pieces)
    Discontinued
    Supplier Number: 0030089774
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89510-210
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 0.2 ml
    Light blue
    100 pieces (4 reclosable bags×25 pieces)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $343.00
    $343.00
    Pack of 100
       $343.00
    Supplier Number: 0030089774
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-308
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 0.5 ml
    Violet
    100 pieces (4 reclosable bags×25 pieces)
    Discontinued
    Supplier Number: 0030089782
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89510-212
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 0.5 ml
    Violet
    100 pieces (4 reclosable bags×25 pieces)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $343.00
    $343.00
    Pack of 100
       $343.00
    Supplier Number: 0030089782
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-310
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 1.0 ml
    Yellow
    100 pieces (4 reclosable bags×25 pieces)
    Discontinued
    Supplier Number: 0030089790
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89510-214
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1.0 ml
    Yellow
    100 pieces (4 reclosable bags×25 pieces)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $343.00
    $343.00
    Pack of 100
       $343.00
    Supplier Number: 0030089790
    Specifications:
    More
    Specifications
    Cat. No.
    CA11033-312
    Environmentally Preferable
    Environmental certification
  • 2.5 ml
    Green
    100 pieces (4 reclosable bags×25 pieces)
    Discontinued
    Supplier Number: 0030089804
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89510-216
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 2.5 ml
    Green
    100 pieces (4 reclosable bags×25 pieces)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $343.00
    $343.00
    Pack of 100
       $343.00
    Supplier Number: 0030089804
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-314
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 5.0 ml
    Blue
    100 pieces (4 reclosable bags×25 pieces)
    Discontinued
    Supplier Number: 0030089812
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89510-218
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 5.0 ml
    Blue
    100 pieces (4 reclosable bags×25 pieces)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $343.00
    $343.00
    Pack of 100
       $343.00
    Supplier Number: 0030089812
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-316
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 10 ml
    Orange
    100 pieces (4 reclosable bags×25 pieces)
    Discontinued
    Supplier Number: 0030089820
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89510-220
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 10 ml
    Orange
    100 pieces (4 reclosable bags×25 pieces)
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $343.00
    $343.00
    Pack of 100
       $343.00
    Supplier Number: 0030089820
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-318
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 25 ml
    Red
    100 pieces (4 reclosable bags×25 pieces) each box containing one adapter
    Discontinued
    Supplier Number: 0030089839
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89510-222
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
    Product available on GSA Advantage®
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 25 ml
    Red
    100 pieces (4 reclosable bags×25 pieces) each box containing one adapter
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $402.00
    $402.00
    Pack of 100
       $402.00
    Supplier Number: 0030089839
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-320
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 50 ml
    Light grey
    100 pieces (4 reclosable bags×25 pieces) each box containing one adapter
    Discontinued
    Supplier Number: 0030089847
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    89511-338
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • 50 ml
    Light grey
    100 pieces (4 reclosable bags×25 pieces) each box containing one adapter
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $402.00
    $402.00
    Pack of 100
       $402.00
    Supplier Number: 0030089847
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    CA11033-322
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable Energy Certificate, ISO 14001, ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure