WYPALL® X80 Wipers, KIMBERLY-CLARK PROFESSIONAL®
Fournisseur: Kimberly-Clark
Certificats
À propos de cet article
Wipers absorb oil and water faster than rags or laundered towels.They maintain strength even in solvents. Long-lasting, on-the-job performance extends usage, reducing cost and waste.
- Durable and economical alternative to laundered shop towels and rags
- Suitable for a wide range of laboratory and industrial tasks, including oil, solvent, and chemical cleanup
- Consistent size and strength without the inconvenience of holes, strings, or residues
- HYDROKNIT® hydroentangled material manufactured without adhesives or chemical binders will not scratch skin or delicate surfaces
- Reusable towels will not fall apart when wet
Ordering information: Quarter-fold packaged wipers are wrapped in stacks of 50. BRAG™ box-packaged wipers are supplied 160 per box; Pop-Up™ boxes each contain 80 wipers.
Frequently Bought Together