ContactUsLinkComponent
Synthware Adapter, Gas Inlet/Outlet, Microscale, Kemtech America
Synthware Adapter, Gas Inlet/Outlet, Microscale, Kemtech America
Catalog # 60004-100
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Adapter, Gas Inlet/Outlet, Microscale, Kemtech America
Catalog # 60004-100
Supplier:  Kemtech America
Supplier Number:  A301410
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • ST Joint:
    14/10
  • Tubing ext.Ø:
    8 mm
  • Cat. no.:
    60004-100
  • Supplier No.:
    A301410
Specifications
About this item
Gas inlet/outlet joint adapter is suitable for microscale applications.
  • For gas inlet/outlet
  • [ST] joint
Adapter is equipped with a standard taper joint.