Synthware Inlet Adapters, Long Stem, Kemtech America
These inlet adapters feature a long stem.
- Top outer [ST] joint
- Bottom inner [ST] joint
- Sealed-in lower tube extending below inner joint
Adapters have top outer and lower inner [ST] joints and sealed-in lower tube extending below inner joint. Tube length is measured below joint.
Frequently Bought Together