Bench Cans, Justrite®
Bench Cans, Justrite®
 :   Justrite
Bench Cans, Justrite®
 :  Justrite
Bench Cans, Justrite®
Bench Cans, Justrite®
Terne plate steel bench can for convenient cleaning of small parts in solvents
Simply place part on spring-loaded, perforated dasher plate, press down and agitate, then release to return dasher to drain position. Plated steel dasher is an effective flame arrester. Hinged cover minimizes evaporation losses. Protected rim reduces possibility of hand injury. Item 56609-724 is a can with parts basket.
Certifications: FM approved.
Product Family Options
  • Capacité
    O.D.×H
  • 0.9 L (0.25 gal.)
    12.3 x 13.9 cm (47/8 x 51/2")
    Each
    Each - $296.87
    $296.87
    Each
       $296.87
     : 10175
  • 1.9 L (0.5 gal.)
    23.8 x 8.25 cm (93/8 x 31/4")
    Each
    Each - $166.24
    $166.24
    Each
       $166.24
     : 10295
  • 3.8 L (1 gal.)
    23.8 x 11.4 cm (93/8 x 41/2")
    Each
    Each - $190.01
    $190.01
    Each
       $190.01
     : 10370
  • 3.8 L (1 gal.)
    23.8 x 11.4 cm (93/8 x 41/2")
    Each
    Each - $168.29
    $168.29
    Each
       $168.29
     : 10375
  • 7.6 L (2 gal.)
    29.2 x 14.9 cm (111/2 x 57/8")
    Each
    Each - $286.83
    $286.83
    Each
       $286.83
     : 10575
  • 11.4 L (3 gal.)
    34.2 x 15.2 cm (131/2 x 6")
    Each
    Each - $300.42
    $300.42
    Each
       $300.42
     : 10775