Nonmetallic Liquid Disposal Cans, Justrite®
Nonmetallic Liquid Disposal Cans, Justrite®
Supplier:   Justrite
Nonmetallic Liquid Disposal Cans, Justrite®
Supplier:  Justrite
Certificates
Nonmetallic Liquid Disposal Cans, Justrite®
Nonmetallic Liquid Disposal Cans, Justrite®
2 Options Available Below
Product Details & Documents
High-density polyethylene cans for containment of corrosives and liquid flammable wastes.
  • High-density polyethylene cans safely contain corrosives or liquid flammable wastes
  • Flame arrester guards against outside ignition
  • Built-in carbon insert provides grounding path
  • Large openings are strategically placed for controlled filling and pouring
  • Fixed-position handle keeps lid open for easier pouring
  • When closed, lid automatically vents at 5psig
  • FM approved
Large openings are strategically placed for controlled filling/pouring during disposal. Fixed-position handle allows lid to stay open for easier pouring. Closed, the cap automatically vents at 5psig. Flame arrester guards against outside ignition. Built-in carbon insert provides a grounding path. Plated steel fittings are ideal for use with flammable liquids. Stainless steel fittings are designed for use with corrosives.
Certifications: FM approved.
Product Family Options
Informations sur le produit
  • Capacité
    O.D.×H
    Availability
    Price
  • 18.9 L (5 gal.)
    30.5x50.8 cm (12x20)"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,101.06
    $1,101.06
    Prix promotionnel pour les nouveaux clients
    Each
       $1,101.06
    Supplier Number: 14765
  • 7.6 L (2 gal.)
    30.5x37.5 cm (12x143/4)"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $950.47
    $950.47
    Prix promotionnel pour les nouveaux clients
    Each
       $950.47
    Supplier Number: 14762
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Stainless steel flame arrester
    Stainless steel flame arrester
    56609-751
    Log in pour la disponibilité
    Each - 482,16 $