Synthware Vacuum Adapters, 1:5 Fluoropolymer Resin Stopcock, Kemtech America
Supplier: Kemtech America
Certificates
About this item
These vacuum adapter feature an inner [ST] joint, fluoropolymer resin stopcock, and 90° hose connection.
- Inner [ST] joint
- Fluoropolymer resin stopcock
- 90° angle hose connection
The stopcock bore is 2mm and the hose connection O.D. is 8mm.
Frequently Bought Together