Nalgene® Safety Half-Liter Bottle Carriers, Thermo Scientific
Catalog # 56609-109
Supplier: Thermo Fisher Scientific
Specifications
- Description:LDPE safety bottle carrier, pint
- Cat. no.:56609-109
- L×W×H:35.6×25.4×27.9 cm (14×10×11")
- Supplier No.:6505-0010
Specifications
About this item
One bottle carrier easily and safely holds up to six standard half-liter reagent bottles in any profile
They can also be used to carry most other half-liter glass or plastic bottles. Bottles are held securely, reducing the possibility they might bump into each other and crack or chip. Each carrier has a sturdy, wide handle molded in one piece with the base, allowing a comfortable grip even when the carrier is full. Snug-fitting cover latches securely to base. Carriers are made of linear, low-density polyethylene and are stackable.