Bottle Jacket, Justrite®
Bottle jacket for PE bottles
Catalog # 56610-100
Supplier: Justrite
Supplier #: 12009
Jump to:
Product Details & Documents
Bottle jacket gives the added protection of a polyethylene bottle without contaminating high purity solvents
Fits 4L and 1gal. bottles. Lab bottles fit to the bottom half of the cover, and lid twists securely for safe pouring. Jacket also acts as a shock absorber, minimizing breakage and spills due to bumps or falls.
Specifications
- Désignation:Bottle jacket for PE bottles