Sontara® AC™ Aircraft Wipers, Contec®
Sontara® AC™ Aircraft Wipers, Contec®
Supplier:   Contec
Sontara® AC™ Aircraft Wipers, Contec®
Supplier:  Contec
Certificates
Sontara® AC™ Aircraft Wipers, Contec®
Sontara® AC™ Aircraft Wipers, Contec®
3 Options Available Below
Product Details & Documents
Sontara® AC™ aircraft wipes leave surfaces clean because they are extremely low-linting, and they tackle tough cleaning agents without falling apart.
  • Low-lint construction is suitable for aircraft manufacturing and maintenance
  • Durable fabric for tough cleaning applications
These are specially engineered to resist snagging on metal parts and corners. That's why they are superior to paper wipes, which snag and tear, leaving lint behind. These can also outperform cheesecloth, which can leave strings behind when it snags on a rivet. Their strength and durability make them ideal for a wide variety of general-purpose and pre-paint applications.
Product Family Options
Informations sur le produit
  • Désignation
    W×L
    Packaging
    Availability
    Price
  • Sontara® AC™ Aircraft Wipes, Quarter-folded
    30.5×33 cm (12×13")
    500/Box, 1 Box/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $202.10
    $202.10
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $202.10
    Supplier Number: AC1213
  • Sontara® AC™ Aircraft Wipes, Pop-Up Box
    30.5×41.9 cm (12×16.5")
    250/Box, 1 Box/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $171.14
    $171.14
    Prix promotionnel pour les nouveaux clients
    Case of 250
       $171.14
    Supplier Number: AC12165
  • Sontara® AC™ Aircraft Wipes, Pop-Up Box
    22.9×41.9 cm (9×16.5")
    100/Box, 8 Boxes/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 800
    Case of 800 - $416.24
    $416.24
    Prix promotionnel pour les nouveaux clients
    Case of 800
       $416.24
    Supplier Number: AC9165