TuffStuff™ Critical Task Wipers, Contec® | Avantor
TuffStuff™ Critical Task Wipers, Contec®
TuffStuff™ Critical Task Wipers, Contec®
Supplier:  CONTEC, INC. MX
Certificates
TuffStuff™ Critical Task Wipers, Contec®
TuffStuff™ Critical Task Wipers, Contec®
4 Options Available Below
Product Details & Documents
TuffStuff™ Wipes are high-quality critical task wipes. Available in medium and heavy-duty formats, these wipes are solvent resistant and maintain their strength even when wet. TuffStuff™ wipes offer exceptional durability and cleanliness.
  • Hydroentangled nonwoven wipes are strong and durable without the use of binders
  • Medium and heavy-duty wipes available to match the ideal wipe for the specific application
  • Offers strength wet or dry, no residues or binders left behind
  • Highly sorbent without the use of surfactants
  • Multiple wipe sizes allow optimum selection for each job
TuffStuff™ Wipes are available in multiple sizes, weights, and packaging configurations to meet various customer needs. The expanded range includes TuffSat, consisting of heavy-duty dry wipes, quarter-folded in a tub that can be saturated with cleaning solution. This versatile option allows wipes to be customized with the correct cleaning solution for a particular environment.
Certifications: Recommended for ISO Class 6 to 8 environments.
Product Family Options
Informations sur le produit
  • Désignation
    W×L
    Packaging
    Availability
    Price
  • TuffStuff™ medium-duty wipes, quarter-folded
    30.5×33 cm (12×13")
    50/Bag, 18 Bags/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 900
    Case of 900 - $258.86
    $258.86
    Prix promotionnel pour les nouveaux clients
    Case of 900
       $258.86
    Supplier Number: TSWL1213
  • TuffStuff™ heavy-duty wipes, quarter-folded
    30.5×43.2 cm (12×17")
    500/Bag, 2 Bags/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $393.88
    $393.88
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $393.88
    Supplier Number: C1C1217Q
  • TuffStuff™ heavy-duty wipes, pop-up dispenser box
    22.9×41.9 cm (9×16.5")
    100/box, 8 boxes/case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 800
    Case of 800 - $329.38
    $329.38
    Prix promotionnel pour les nouveaux clients
    Case of 800
       $329.38
    Supplier Number: TSWC0917
  • TuffStuff™ heavy-duty wipes, double pop-up dispenser box
    30.5×41.9 cm (12×16.5")
    125/Box, 2 Boxes/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 250
    Case of 250 - $123.84
    $123.84
    Prix promotionnel pour les nouveaux clients
    Case of 250
       $123.84
    Supplier Number: TSWC1217