IsoTherm-System® Sample Cooling System | Avantor
IsoTherm-System® Sample Cooling System
Supplier:   Eppendorf
IsoTherm-System® Sample Cooling System
Supplier:  Eppendorf
Certificates
IsoTherm-System® Sample Cooling System
IsoTherm-System® Sample Cooling System
3 Options Available Below
Product Details & Documents
Cool samples safely and efficiently using the Eppendorf® IsoTherm-System. Choose between IsoPacks for 0 °C and –21 °C and use the IsoRack for centrifuging, transporting, or storing, ensuring constant temperature maintenance throughout your workflow.
  • Efficiently cool samples at a constant temperature and then thaw them gently
  • IsoRack suitable for centrifuging in a plate rotor with IsoRack adapter
  • IsoRack serves as a work rack when filling micro test tubes and for provisioning, sorting, distributing, transporting, storing, or freezing samples in micro test tubes
  • Bore holes at the distances of the MTP grid, allow for micro test tubes to be filled using multi-channel pipettes
IsoRack: autoclavable at 121 °C, 20 min; IsoPack and IsoSafe: not autoclavable.
Temperature Resistance: IsoRack: –200 to 100 °C.
IsoRack: polycarbonate; IsoPack: polyethylene, bottom: aluminum foil; IsoSafe: polypropylene.
Store dry at room temperature. Protect from direct sunlight and UV light. IsoRack: UV resistant.
Max. 2250 ×g (IsoRack in IsoRack adapter).
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Packaging
    Availability
    Price
  • IsoTherm-System® starter set, 0 °C and –21 °C temperature control
    24 micro test tubes (1.5/2.0 ml)
    Includes IsoSafe, IsoRack, 0 °C IsoPack, and –21 °C IsoPack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $377.00
    $377.00
    Each
       $377.00
    Supplier Number: 022510053
  • IsoPack and IsoRack set, 0 °C temperature control
    24 micro test tubes (1.5/2.0 ml)
    1×IsoPack and 1×IsoRack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.00
    $119.00
    Each
       $119.00
    Supplier Number: 022510240
  • IsoPack and IsoRack set, –21 °C temperature control
    24 micro test tubes (1.5/2.0 ml)
    1×IsoPack and 1×IsoRack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.00
    $119.00
    Each
       $119.00
    Supplier Number: 022510258