PhoneWipe® Presaturated Pad, CleanTex™
14 x 20.3 cm (51/2 x 8")
Catalog # 58019-508
Supplier: Advantus
: CT808
Jump to:
Product Details & Documents
For cleaning and disinfecting telephones, microphones, and other shared communication equipment.
This disposable HydroSorb® (cellulose/polyester) pad is presaturated with benzethonium chloride disinfectant that kills most germs on hard, nonporous surfaces. Each pad is hermetically sealed and individually wrapped.
Spécifications
- W×L:14 x 20.3 cm (51/2 x 8")