PYREX® Roller Bottles, Corning
 :   Corning
PYREX® Roller Bottles, Corning
 :  Corning
PYREX® Roller Bottles, Corning
PYREX® Roller Bottles, Corning
PYREX®, borosilicate glass.
  • Optical clarity and mechanical strength
  • Withstand wet or dry sterilization
 : Supplied with 38 mm deep skirted GPI 38-430 threaded phenolic screw cap with rubber liner.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
  • Catalog No:
  • 22877-218
  • 22877-228
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Product Family Options
  • Désignation
    Culture Area
    Outer Diameter
    Taille du bouchon
    Cap Type
    Hauteur
    Medium Volume
  • Straight Neck, Threaded
    840 cm²
    108 mm
    38 mm
    PP cap
    204 mm
    168 - 252 ml
    Case of 2
    Case of 2 - $423.00
    $423.00
    Case of 2
       $423.00
     : 1400-285
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    22877-218
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Short Neck, Threaded
    840 cm²
    110 mm
    45 mm
    PP cap
    285 mm
    168 - 252 ml
    Case of 2
    Case of 2 - $583.00
    $583.00
    Case of 2
       $583.00
     : 1425-285
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    22877-228
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact