Premoistened polyester wipes are designed for general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments.
- Ideal for general purpose and critical cleaning
- Wipes are chemical resistant and exceptionally low in particles and extractable residue
- Presaturation greatly reduces solvent usage
Standard weight wipes are made of 100% textured polyester with no-run interlock knit for good solvent sorbency, abrasion resistance, and chemical resistance. Heatsealed and saturated with various blends of isopropyl alcohol and deionized water. Low in particles and extractables. Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Sterile wipers are Validated Sterile to 10⁻⁶ Sterility Assurance level in accordance with AAMI/ISO 11137 Method 1 Guidelines for the sterilization of medical devices. Sterile PROSAT® Knitted Polynit Wipes are conveniently packaged for use in sterile core areas and are ideal for wiping articles prior to pass-through and environmental cleaning.
Ideal for wiping exterior product surfaces and cleaning the interior surfaces of process tools or other equipment.