SATWipes® Sigma™ Wipes, Contec®
Supplier: Contec
Certificates
About this item
The SATWIPES® Sigma™ consists of a perforated roll of saturated wipes which are dispensed through a center-pull canister. The wipes are made of 55% cellulose and 45% polyester creating highly sorbent and durable wipe.
- Hydraulically entangled fabric creates a soft yet durable wipe
- Clean wipe offers consistent solvent application
- Low levels of particles and extractable contaminants to reduce contamination possibility
The isopropanol solvent blends provide safe cleaning solutions in various concentrations. Packaged with refill packs and a reusable pop-up dispenser, eliminating the need for solvent bottles and reducing solvent handling.
Frequently Bought Together