Oily Waste Cans, Justrite®
Supplier: Justrite
Certificates
About this item
Essential wherever solvent cloths and wiping rags are used
- Can be operated by hand or foot-lever opening device
- Lid closes automatically when not in use
- Round construction and elevated bottom encourage circulation of air
- One-piece cover facilitates use of collection bags
- Steel construction with powder coat paint finish
When operated by hand or foot lever opening device, the cover opens to a 60° angle and closes automatically when released. Round construction and elevated bottom encourage circulation of air around can to disperse heat and prevent spontaneous combustion. One-piece cover facilitates use of collection bags. Steel construction. Powder coat paint finish. UL listed. FM approved.
Can remains closed when not in use.
Frequently Bought Together