Synthware Gas Washing Bottle with Fritted Disc and Two Hose Connections, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Gas washing bottle with [ST]45/50 cap-style stopper and fritted disc angled at 15° for even distribution of gas throughout absorbing material.
- Heavy wall borosilicate construction
- Fritted disc
- Large base
- 45/50 ground glass joint
Available with a coarse or extra-coarse fritted disc. Springs (60000-758) are ordered separately.
Frequently Bought Together