- Description:Wet & Dry™ Twinpaks
- Cat. no.:58019-514
- Supplier No.:CT812
Use this system for cleaning, drying and controlling static on terminal, CRT and COM screens, display screens and instrument glass.
Not for use on nylon mesh antiglare screens.
Two pads are hermetically sealed in separate foil packets and attached in the middle. The wet pad is moistened with a special antistatic screen cleaning solution (ultrafiltered to 0.2 µl) that neutralizes electrostatic charges while removing contaminants from the screen. Made from 100% lint-free polyester spunbonded fabric that contains no binders or additives to affect surface being cleaned. The dry pad is a pure, nonwoven cloth combining the absorbency of cellulose with the strength of polyester. This high absorbency drying pad polishes screens to a streak-free finish.