Biohazard Waste Containers, Justrite® | Avantor
Biohazard Waste Containers, Justrite®
Supplier:   Justrite
Biohazard Waste Containers, Justrite®
Supplier:  Justrite
Certificates
Biohazard Waste Containers, Justrite®
Biohazard Waste Containers, Justrite®
2 Options Available Below
Product Details & Documents
For temporary collection of hazardous biological waste
  • For segregation of infectious waste
  • Leakproof, sturdy metal construction
  • Simple, foot-operated, self-closing lid to reduce contamination
  • Biohazard bag is held securely around the circumference of the can
Caution: Not to be used with sharps.
May be used for temporary storage of contaminated laundry as well as other regulated waste.
Product Family Options
Informations sur le produit
  • Capacité
    Dimensions
    Availability
    Price
  • 22.7 L (6 gal.)
    30.1O.D x 40.3H cm (117/8 x 157/8")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $494.89
    $494.89
    Prix promotionnel pour les nouveaux clients
    Each
       $494.89
    Supplier Number: 05910
  • 37.9 L (10 gal.)
    35.3O.D x 45.7H cm (1315/16 x 18")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $664.56
    $664.56
    Prix promotionnel pour les nouveaux clients
    Each
       $664.56
    Supplier Number: 05930
Accessories
Informations sur le produit
  • Catalog #
    Color
    Capacity
    Availability
    Price
  • Biohazard bags
    Biohazard bags
    56617-834
    Red
    15 gal.
    Log in pour la disponibilité
    Pack of 100 - $264.13