Terminal® Biohazard Benchtop Keeper™ Containers, Whitney Products
Fournisseur: Heathrow Scientific
Certificats
À propos de cet article
Use large or small containers for the collection of biohazardous items such as pipet tips, petri dishes, test tubes, and slides.
- For collection of biohazardous items such as pipet tips, petri dishes, test tubes, and slides
- Sturdy, plastic-lined paperboard construction
- Leak-resistant and autoclavable
Constructed of sturdy, plastic-lined paperboard. Disposable containers are leak-resistant and autoclavable. Self-locking top for safe closure.
Frequently Bought Together