Separatory Funnels, Pear-Shape, Glas-Col® | Avantor
Separatory Funnels, Pear-Shape, Glas-Col®
Supplier:   Glas-Col
Separatory Funnels, Pear-Shape, Glas-Col®
Supplier:  Glas-Col
Certificates
Separatory Funnels, Pear-Shape, Glas-Col®
Separatory Funnels, Pear-Shape, Glas-Col®
4 Options Available Below
Product Details & Documents
Funnels have a screw cap with a fluoropolymer resin lining to reduce the possibility of contamination
Pressure release valves offer the convenience of self-venting for applications where pressure build-up is a concern. Each valve is made of fluoropolymer resin and is factory preset to release at 1psi of pressure. The valve can also be adjusted to release at pressures up to 5psi by turning the pressure release slug on the top of the valve.
The drain spout is a screw-on fluoropolymer resin design, which reduces the chance of breakage and allows for easier cleaning.
Product Family Options
Product Information
  • Désignation
    Capacité
    Availability
    Price
  • Funnel
    250 ml
    Log in for availability
    Log in for availability
    Each
    Each - $611.66
    $611.66
    Each
       $611.66
    Supplier Number: 099A SEP0250
  • Funnel
    500 ml
    Log in for availability
    Log in for availability
    Each
    Each - $850.87
    $850.87
    Each
       $850.87
    Supplier Number: 099A SEP0500
  • Funnel
    1000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $1,022.21
    $1,022.21
    Each
       $1,022.21
    Supplier Number: 099A SEP1000
  • Funnel
    2000 ml
    Log in for availability
    Log in for availability
    Each
    Each - $1,230.94
    $1,230.94
    Each
       $1,230.94
    Supplier Number: 099A SEP2000
Accessories
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Pressure Release Valve
    Pressure Release Valve
    80078-774
    For 60 - 1000 ml funnels
    Log in for availability
    Each - $556.73
Product Information
  • Catalog #
    Capacity
    Availability
    Price
  • Pressure Release Valve
    Pressure Release Valve
    80078-776
    For 2000 ml funnels
    Log in for availability
    Each - $556.73