Diversified Biotech CoolSafe™ Systems
Supplier:   Diversified Biotech
Diversified Biotech CoolSafe™ Systems
Certificates
Diversified Biotech CoolSafe™ Systems
Diversified Biotech CoolSafe™ Systems
8 Options Available Below
Product Details & Documents
Anodized aluminum storage kits are ideal for sustaining cool or warm constant temperatures up to 10 hours during benchtop storage
Each kit includes one polystyrene CoolSafe storage box, one anodized aluminum cooling chamber rack, and two interchangeable cool bricks. Additional aluminum cooling chamber racks are also available.
Product Family Options
Storage Kits for Plates
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • CoolSafe System for 384-Well Plates
    For 384-Well Plates
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,624.43
    $2,624.43
    Each
       $2,624.43
    Supplier Number: CSAF-3840
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-868
    Small Business Enterprise
  • CoolSafe System for Flat-Bottom Microplates
    For Flat-Bottom Microplates
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,872.74
    $1,872.74
    Each
       $1,872.74
    Supplier Number: CSAF-9600
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-874
    Small Business Enterprise
Storage Kits for Tubes
Informations sur le produit
  • Désignation
    Capacité
    Availability
    Price
  • CoolSafe System for 0.2 mL Tubes (Also fits PCR Plates)
    For PCR Plates or 0.2 mL Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,174.57
    $2,174.57
    Each
       $2,174.57
    Supplier Number: CSAF-1000
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-860
    Small Business Enterprise
  • CoolSafe System for 0.5 mL Tubes
    For 0.5 mL Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,097.97
    $2,097.97
    Each
       $2,097.97
    Supplier Number: CSAF-2000
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-862
    Small Business Enterprise
  • CoolSafe System for 1.5 mL Tubes
    For 1.5 mL Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,827.25
    $1,827.25
    Each
       $1,827.25
    Supplier Number: CSAF-3000
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-864
    Small Business Enterprise
  • CoolSafe System for 1.5 mL Cryo Tubes
    For 1.5 mL Cryogenic Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,827.28
    $1,827.28
    Each
       $1,827.28
    Supplier Number: CSAF-3050
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-866
    Small Business Enterprise
  • CoolSafe System for 5 mL Tubes
    For 5 mL Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,994.92
    $1,994.92
    Each
       $1,994.92
    Supplier Number: CSAF-4000
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-870
    Small Business Enterprise
  • CoolSafe System for 0.2 mL, 0.5 mL and 1.5 mL Tubes
    For 0.2 mL, 0.5 mL, and 1.5 mL Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,885.51
    $1,885.51
    Each
       $1,885.51
    Supplier Number: CSAF-5000
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89024-872
    Small Business Enterprise
Accessories
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-880
    Holds Forty-Eight 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,301.34
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-882
    Holds Fifty-Five 1.5 mL Cryogenic Tubes
    Log in pour la disponibilité
    Each - $1,289.09
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-884
    Holds One 384-Well Plate, Six 0.5 mL Tubes, and Four 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $2,268.99
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-878
    Holds Ninety-One 0.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,752.18
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-886
    Holds Thirty-Two 5 mL Tubes
    Log in pour la disponibilité
    Each - $1,449.35
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-876
    Holds One PCR Plate or Ninety-Six 0.2 mL Tubes, Six 0.5 mL Tubes, and Four 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,462.18
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-888
    Holds Twenty-Four 0.2 mL Tubes, Thirty-Five 0.5 mL Tubes, and Twenty 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,396.85
Informations sur le produit
  • Catalog #
    Details
    Availability
    Price
  • Replacement Cooling Chamber Racks
    Replacement Cooling Chamber Racks
    89024-890
    Holds One Flat-Bottom Microplate, Six 0.5 mL Tubes, and Four 1.5 mL Tubes
    Log in pour la disponibilité
    Each - $1,326.60