Olympic™ Stacking Bins, Metro™
Olympic™ Stacking Bins, Metro™
Supplier:   Metropolitan Wire
Olympic™ Stacking Bins, Metro™
Supplier:  Metropolitan Wire
Certificates
Olympic™ Stacking Bins, Metro™
Olympic™ Stacking Bins, Metro™
5 Options Available Below
Product Details & Documents
Bins are durable, stackable, and feature high impact strength
  • A flat surface on the front of the bin is provided for easy placement of labels and bar codes
  • All bins have an open hopper for easy access to contents
  • Bins feature a lip on the edge of the rear panel for hanging
Choose from Benstat™ or Bentron™ bins:
Benstat is a static-dissipative polypropylene-based material with a surface resistivity of >105ohms/sq. and <1012ohms/sq. All Benstat bins are blue.
Bentron is a polypropylene-based material with a carbon additive to provide an electrically conductive path through the material. Bentron has a surface resistivity of <105ohms/sq. All Bentron bins are black.
Product Family Options
Benstat™ Blue Static-Dissipative Stacking Bins
Product Information
  • Désignation
    Color
    Dimensions
    Availability
    Price
  • Benstat bin
    Blue
    14W x 27.6L x 12.7H cm (51/2 x 107/8 x 5")
    Log in for availability
    Log in for availability
    Pack of 24
    Pack of 24 - $1,223.92
    $1,223.92
    Pack of 24
       $1,223.92
    Supplier Number: SB91055BAS
  • Benstat bin
    Blue
    10.5W x 18.8L x 7.6H cm (41/8 x 73/8 x 3")
    Log in for availability
    Log in for availability
    Pack of 48
    Pack of 48 - $1,210.55
    $1,210.55
    Pack of 48
       $1,210.55
    Supplier Number: SB90743BAS
  • Benstat bin
    Blue
    10.5W x 18.8L x 7.6H cm (41/8 x 73/8 x 3")
    Discontinued
    Supplier Number: -
  • Benstat bin
    Blue
    14W x 27.6L x 12.7H cm (51/2 x 107/8 x 5")
    Discontinued
    Supplier Number: -
Bentron™ Black Conductive Stacking Bins
Product Information
  • Désignation
    Color
    Dimensions
    Availability
    Price
  • Bentron bin
    Black
    10.5W x 18.8L x 7.6H cm (41/8 x 73/8 x 3")
    Log in for availability
    Log in for availability
    Each
    Each - $27.64
    $27.64
    Each
       $27.64
    Supplier Number: SB90743CAS
  • Bentron bin
    Black
    14W x 27.6L x 12.7H cm (51/2 x 107/8 x 5")
    Log in for availability
    Log in for availability
    Each
    Each - $55.58
    $55.58
    Each
       $55.58
    Supplier Number: SB91055CAS
  • Bentron bin
    Black
    21W x 37.5L x 17.8H cm (81/4 x 143/4 x 7")
    Discontinued
    Supplier Number: -
  • Bentron bin
    Black
    41.9W x 37.5L x 17.8H cm (81/4 x 161/2 x 7")
    Log in for availability
    Log in for availability
    Each
    Each - $119.79
    $119.79
    Each
       $119.79
    Supplier Number: SB91516CAS