About this item
These pillow-style mop heads move easily up and down and from side to side, ideal for precise cleaning of shelves, narrow chambers, small spaces and for manual wall wiping. Designed for use with the PilloMop™ adapter and SSU series Universal Handles.
- Mop heads easily attach to the acetal plate mop frame via velcro strips
- Adapter features 360° swivel to easily reach awkward corners and angles
- Available in polyester or PVA fabric covers with urethane foam interiors
- Single-ply slipcover option for stand alone use or over padded mop
- Compatible with IPA, bleach, peroxide and string disinfectants and cleaning solutions
- Adapter attaches to SSU series handles via Quick Disconnect attachment
- Available gamma irradiated for aseptic environments
- System is autoclavable, reusable and can be laundered
The Micronova PilloMop™ easily attaches to the sturdy acetal plate PilloMop™ Adapter via two polyester, hook and loop (velcro) strips. The NovaPoly™ PilloMop™ is low-linting for critical and sterile applications. The PVA PilloMop™ is designed for semiconductor, electronics, aerospace and other applications requiring low-linting absorbance in critical cleaning protocols. A single layer polyester slipcover without foam interior (PMC-648) may be used alone or over the pillow-style mops and can be changed with each wipe, offering precise manual wall wiping control.
Specifications
- Catalog No:
- 68400-238
- 43300-956
- Environmentally Preferable:
- Waste Reducing