PolyGen™ Mops, Micronova | Avantor
PolyGen™ Mops, Micronova
   Options durables disponibles
Supplier:   Micronova
PolyGen™ Mops, Micronova
Supplier:  Micronova
   Options durables disponibles
Certificates
PolyGen™ Mops, Micronova
3 Options Available Below
Product Details & Documents
PolyGen™ mops are ideal for disinfecting in pharmaceutical and biotech environments.
  • Low-linting polyester looped strands with dual-action abrasive mesh
  • Herringbone weave ensures there are no raw edges, and limits particle generation
  • Mops are reuseable, launderable, and autoclavable
LoopMop™ mops are knitted polyester looped mops for applying disinfectant in pharmaceutical, aseptic processing, and medical device applications. The polyester material is absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants that require an extended drying time. LoopMop™ mops are reuseable, launderable, and autoclavable.
All mop heads attach to the stainless steel universal handle by means of the orange polypropylene quick-disconnect adapter. For less critical applications, choose the fiberglass/polypropylene handle with attached adapter (68400-002).
Ordering information: All components are ordered separately. For additional handle lengths, see the 68400-302 series.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 68400-786
  • 68399-706
  • 68400-758
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Sterility
    Availability
    Price
  • LoopMop™ Mop Head
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $2,640.00
    $2,640.00
    Case of 24
       $2,640.00
    Supplier Number: MZLM5-12IR
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    68400-786
    Environmentally Preferable
    Waste Reducing
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
  • PolyGen™ Mop Head
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $104.00
    $104.00
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,124.52
    $1,124.52
    Each
       $104.00
    Case of 12
       $1,124.52
    Supplier Number: PG9-12
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    68399-706
    Environmentally Preferable
    Waste Reducing
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
  • LoopMop™ Mop Head
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $72.10
    $72.10
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $1,626.24
    $1,626.24
    Each
       $72.10
    Case of 24
       $1,626.24
    Supplier Number: MZLM5-12
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    68400-758
    Environmentally Preferable
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
Accessories
Informations sur le produit
  • Catalog #
    Material
    Length
    Availability
    Price
  • MicroMop™ adapter and handle
    MicroMop™ adapter and handle
    68400-002
    Polypropylene adapter, fiberglass handle
    182.9 cm (72")
    Log in pour la disponibilité
    Each - $92.00
Informations sur le produit
  • Catalog #
    Material
    Length
    Availability
    Price
  • MicroMop™ adapter, attaches to SSU series handles
    MicroMop™ adapter, attaches to SSU series handles
    68400-202
    Stainless steel, 304 electropolished, polypropylene
    -
    Log in pour la disponibilité
    Each - $159.00
Informations sur le produit
  • Catalog #
    Material
    Length
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Stainless steel quick disconnect extendable handle
    68400-164
    Stainless steel 304, electropolished
    Extendable from 96.5 - 182.9 cm (38 - 72")
    Log in pour la disponibilité
    Each - $464.98