Slimline™ Flathead Mop Covers and Hardware, Micronova
A series of elasticized slipcovers, with urethane foam interiors, that are ideal for fine cleaning and sanitizing all surfaces and equipment in the cleanroom. Available without a foam interior or gamma irradiated for aseptic environments.
- Covers are easy to change and fit securely over mop frame
- Fully encased Urethane Foam layer for ample disinfectant dispersal
- Single-ply covers available without foam interior
- Fits on QPSL plastic frame or QDSL stainless steel frame
- Suitable for use with strong disinfectants and repeated autoclaving
The Micronova Slimline™ Flat Head Mop Covers fit easily and securely onto the 360° swivel SlimLine™ plastic or stainless steel mop adapters, which can be locked at 180° for more controlled cleaning.
- Catalog No:
- 43300-966
- 76323-720
- 43300-970
- 75845-734
- 43300-968
- 11310-604
- 11310-602
- 94023-842
- 11310-606
- CA76006-322
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
SlimLine™ Mop Adapters
Frequently Bought Together