VWR® CryoCooler Freeze Controller | Avantor
VWR® CryoCooler Freeze Controller
VWR® CryoCooler Freeze Controller
Catalog # 414004-284
Supplier:   VWR International
VWR® CryoCooler Freeze Controller
Catalog # 414004-284
Supplier:  VWR International
VWR® CryoCooler Freeze Controller
VWR® CryoCooler Freeze Controller
VWR® CryoCooler Freeze Controller
Price
$150.29
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each Case of 8 Pallet of 384
Product Details & Documents
When filled with isopropyl alcohol and placed in mechanical freezer, this polycarbonate freeze controller provides the -1°C per minute cooling rate required to successfully preserve cells.
  • Non-mechanical
  • Successfully preserves cells
  • Insert helps prevent contamination
Floating rigid insert helps prevent contamination. Withstands repeated use. Holds up to eighteen 1.2–2 mL cryogenic vials.
Spécifications
  • Désignation:
    VWR® CryoCooler Freeze Controller