VWR® CryoCoolers
VWR® CryoCoolers
VWR® CryoCoolers
Supplier:  VWR International
Certificates
VWR® CryoCoolers
VWR® CryoCoolers
3 Options Available Below
Product Details & Documents
Coolers hold 0.5 or 1.5 mL microcentrifuge tubes at or below -20°C (-4°F) for up to one hour on the benchtop or in transport.
  • Printed grid
  • Keyed lid
  • Nonskid feet
Cooler and lid are filled with nontoxic gel. Store at -25°C (-13°F).
Parts # 414004-287 and 414004-285 come with a locking handle.
Documents
Product Family Options
Product Information
  • Capacité
    L×W×D
    Availability
    Price
  • 12 Vials
    15.2 x 10.2 x 10.2 cm (6 x 4 x 4")
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $223.75
    $223.75
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $1,790.03
    $1,790.03
    New Customer Promotional Price
    Log in for availability
    Pallet of 224
    Pallet of 224 - $50,120.85
    $50,120.85
    New Customer Promotional Price
    Each
       $223.75
    Case of 8
       $1,790.03
    Pallet of 224
       $50,120.85
  • 20 Vials
    21.6 x 16.5 x 11.4 cm (81/2 x 61/2 x 41/2")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $382.07
    $382.07
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $3,056.56
    $3,056.56
    New Customer Promotional Price
    Each
       $382.07
    Case of 8
       $3,056.56
  • 32 Vials
    22.9 x 14 x 11.4 cm (9 x 51/2 x 41/2")
    Log in for availability
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $445.35
    $445.35
    New Customer Promotional Price
    Log in for availability
    Case of 8
    Case of 8 - $3,562.83
    $3,562.83
    New Customer Promotional Price
    Log in for availability
    Pallet of 96
    Pallet of 96 - $42,754.00
    $42,754.00
    New Customer Promotional Price
    Each
       $445.35
    Case of 8
       $3,562.83
    Pallet of 96
       $42,754.00