CoolSafe Centrifuge Tube Cooling Chambers, Diversified Biotech
Supplier: Diversified Biotech
Certificates
About this item
Cooling chambers are constructed from anodized aluminum blocks and are excellent temperature conductors
Ideal for use when storing samples in ice buckets, freezers, and on hot plates. Chamber 89024-898 features a raised step design for easier viewing of samples.
Frequently Bought Together