MicroMop™ Flat String Mop Heads, Micronova
MicroMop™ Flat String Mop Heads, Micronova
   Options durables disponibles
Supplier:   Micronova
MicroMop™ Flat String Mop Heads, Micronova
Supplier:  Micronova
   Options durables disponibles
Certificates
MicroMop™ Flat String Mop Heads, Micronova
5 Options Available Below
Product Details & Documents
A selection of highly effective, flat-strand mops with high absorbency and low particulates. Available in three materials for specialized cleaning; polyvinyl alcohol, synthetic chamois, and die-cut polyester.
  • High absorbency and low particulates
  • Dry, residue-free cleaning results
  • Designed for cleanroom and controlled environments
Fabric options include:
PVA - non-woven polyvinyl alcohol with 600% absorbency and 10-12 second dry time; ideal for semiconductor and microelectronic cleaning applications
NovaLite™ - absorbent synthetic chamois; excellent fine clean and finish for medical device applications
NovaPoly™ - 100% smooth knit polyester provides excellent disinfectant dispersal for cleanroom sanitization capabilities.
The Micronova MicroMop™ Flat String Mop Heads are designed for use with the Micronova MicroMop™ series hardware: MH-1 or MHC-1 frames with attached handles, or QDMH-1 frame with SSU series handles.
Ordering information: All components must be ordered separately. For additional handle lengths, see Universal Mop Handles from Micronova.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 94023-782
  • 68400-002
  • 68400-202
  • 68400-210
  • 68400-778
  • 68400-001
  • Environmentally Preferable:
  • Waste Reducing
  • Waste Reducing
  • Waste Reducing
Spécifications
Product Family Options
Hardware
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Color
    Availability
    Price
  • Clamp mop adapter and handle
    Polypropylene adapter, fiberglass handle
    152.4 cm (60")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $99.40
    $99.40
    Each
       $99.40
    Supplier Number: MHC-1
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    94023-782
    Environmentally Preferable
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
Handle
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Color
    Availability
    Price
  • MicroMop™ adapter and handle
    Polypropylene adapter, fiberglass handle
    182.9 cm (72")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $92.00
    $92.00
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,432.32
    $1,432.32
    Each
       $92.00
    Case of 12
       $1,432.32
    Supplier Number: MH-1
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    68400-002
    Environmentally Preferable
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
  • MicroMop™ adapter, attaches to SSU series handles
    Stainless steel, 304 electropolished, polypropylene
    -
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $159.00
    $159.00
    Each
       $159.00
    Supplier Number: QDMH-1
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    68400-202
    Environmentally Preferable
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
Mop Heads
Informations sur le produit
  • Désignation
    Matière
    Longueur
    Color
    Availability
    Price
  • MicroMop™ flat strand mop, NovaLite™
    NovaLite™, synthetic chamois
    Orange
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $212.00
    $212.00
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $3,908.28
    $3,908.28
    Each
       $212.00
    Case of 12
       $3,908.28
    Supplier Number: CRM-2
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    68400-210
    Environmentally Preferable
    Waste Reducing
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
  • MicroMop™ flat strand mop, NovaPoly™
    NovaPoly™, 100% smooth knit polyester, die-cut
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $90.70
    $90.70
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $1,023.60
    $1,023.60
    Each
       $90.70
    Case of 12
       $1,023.60
    Supplier Number: CRM-66
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    68400-778
    Environmentally Preferable
    Waste Reducing
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
  • MicroMop™ flat strand mop, PVA
    PVA, non-woven polyvinyl alcohol
    Yellow
    Discontinued
    Supplier Number: -
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
    More
    Specifications
    Cat. No.
    68400-001
    Environmentally Preferable
    Waste Reducing
    Minority or Woman-Owned Business Enterprise
    Small Business Enterprise
      Waste Reducing
Handle
Informations sur le produit
  • Catalog #
    Désignation
    Longueur
    Matière
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Extendable from 96.5 - 182.9 cm (38 - 72")
    Stainless steel 304, electropolished
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $464.98
    $464.98
    Each
       $464.98