Weitlaner Retractors, Sharp, Sklar®
 :   Sklar
Weitlaner Retractors, Sharp, Sklar®
 :  Sklar
Weitlaner Retractors, Sharp, Sklar®
Weitlaner Retractors, Sharp, Sklar®
Sklar's® Weitlaner Retractor is a self-retaining, finger ring retractor with a cam ratchet lock used for holding back tissue and exposing a surgical site. It has curved shanks that lead to blades that have between two and six, sharp or blunt, outward curving prongs. The prongs interdigitate when the instrument is closed.
The Weitlaner retractor is a popular instrument, most commonly used in basic plastic surgery, large bone and joint procedures, and mastoid surgeries
Censitrac Ready products are industry standard and constructed of stainless steel.
It is also useful with small, deep incisions and soft tissue dissection at a superficial level.
 : Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
 : Customers must have a Censitrac Site ID to purchase Censitrac instruments.
  • Catalog No:
  • 82030-142
  • 82030-224
  • 82030-226
  • 82030-228
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex/Latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
Product Family Options
Weitlaner Retractors, Sharp
  • Désignation
    Longueur
    Grade
    Matière
    Curvature
    Handle
    Sterility
    Tip Configuration
    Working Surface Style
  • Weitlaner Self-Retaining Retractor
    11.4 cm (4½")
    Premium OR-Grade
    Stainless steel
    Curved
    Finger Rings
    Non sterile
    2 x 3 Prongs Sharp
    Smooth
    Each
    Each - $838.57
    $838.57
    Each
       $838.57
     : 22-8245
    Specifications
    Cat. No.
    82030-142
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
  • Weitlaner Self-Retaining Retractor
    16.5 cm (6½")
    Premium OR-Grade
    Stainless steel
    Non sterile
    3 x 4 Prongs Sharp
    Each
    Each - $887.42
    $887.42
    Each
       $887.42
     : 42-1965
    Specifications
    Cat. No.
    82030-224
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
  • Weitlaner Self-Retaining Retractor
    20.3 cm (8")
    Premium OR-Grade
    Stainless steel
    Non sterile
    3 x 4 Prongs Sharp
    Each
    Each - $992.53
    $992.53
    Each
       $992.53
     : 42-1975
    Specifications
    Cat. No.
    82030-226
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free
  • Weitlaner Self-Retaining Retractor
    24.1 cm (9½")
    Premium OR-Grade
    Stainless steel
    Non sterile
    3 x 4 Prongs Sharp
    Each
    Each - $1,028.94
    $1,028.94
    Each
       $1,028.94
     : 42-1995
    Specifications
    Cat. No.
    82030-228
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex-free