Sample Vials for Vi-CELL BLU Analyzers
Sample vials for Vi-CELL BLU analyzers
: C99606
Bag contains 500 sample vials for use with the Vi-CELL BLU Analyzer.
- Minimum volume required in sample vial is 200 µm
- In FAST mode, a minimum volume of 170 µm is required in the sample vial
- Désignation:Sample vials for Vi-CELL BLU analyzers